missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals
£23.00 - £364.00
Chemical Identifiers
CAS | 109425-55-0 |
---|---|
Molecular Formula | C25H30N2O6 |
Molecular Weight (g/mol) | 454.523 |
MDL Number | MFCD00065668 |
InChI Key | JOOIZTMAHNLNHE-NRFANRHFSA-N |
Synonym | fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine |
PubChem CID | 2756114 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
SMILES | CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489278
|
Thermo Scientific Alfa Aesar
H62814.03 |
1 g | N/A | ||||||
15499278
|
Thermo Scientific Alfa Aesar
H62814.06 |
5 g |
£86.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409288
|
Thermo Scientific Alfa Aesar
H62814.14 |
25 g |
£364.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109425-55-0 | |
454.523 | |
JOOIZTMAHNLNHE-NRFANRHFSA-N | |
2756114 | |
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C25H30N2O6 | |
MFCD00065668 | |
fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
111°C to 115°C | |
C25H30N2O6 | |
4772025 | |
JOOIZTMAHNLNHE-NRFANRHFSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
2756114 | |
96% |
109425-55-0 | |
MFCD00065668 | |
fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine | |
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
454.523 | |
454.52 | |
Ndelta-Boc-Nalpha-Fmoc-L-ornithine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only