missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Neck Cord
35.40 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Color:
Black
Specifications
Color | Black |
Product Type | Neck Cord |
For Use With (Application) | Holding Spectacles of Universal Sizes |
No. per Pack | 50/Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'