missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nectin-1/PVRL1 Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
£347.00 - £834.00
Specifications
Assay Range | 5000 pg/mL |
---|---|
Assay Sensitivity | 78.125-5000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18282517
|
Novus Biologicals™
NBP2-79660-15PLATES |
15 Plates |
£834.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18278277
|
Novus Biologicals™
NBP2-79660-5PLATES |
5 Plates |
£347.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Nectin-1/PVRL1.Specifications
5000 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Nectin-1/PVRL1. | |
Sandwich ELISA | |
5818 | |
Mouse | |
RUO | |
Nectin-1/PVRL1 |
78.125-5000 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12 | |
NECTIN1 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 78.125 pg/mL is recommended. |