missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nectin-1/PVRL1 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Brand:  Novus Biologicals™ NBP2-79640-5Plates

 View more versions of this product

Product Code. 18603321

  • £341.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Nectin-1/PVRL1.
Specifications

Specifications

23.45 pg/mL
Sandwich ELISA
Sandwich ELISA Kit
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Nectin-1/PVRL1.
Sandwich ELISA
5818
Rabbit
5 Plates
Storage is content dependent.
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
23.45-1500 pg/mL
HRP
Antibody Pairs
Rat
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
NECTIN1
  • Standard: Each vial contains 105 ng of recombinant Rat Nectin-1/PVRL1.
  • Capture Antibody: 2 mg/mL of rabbit anti-Rat Nectin-1/PVRL1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Nectin-1/PVRL1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • RUO
    Nectin-1/PVRL1
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers