missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nectin-1/PVRL1 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

£341.00 - £844.00

Specifications

Assay Range 23.45 pg/mL
Assay Sensitivity 23.45-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18228656
View Documents
Novus Biologicals™
NBP2-79640-15PLATES
15 Plates
£844.00
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
18603321
View Documents
Novus Biologicals™
NBP2-79640-5Plates
5 Plates
£341.00
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Nectin-1/PVRL1.
Specifications

Specifications

23.45 pg/mL
Sandwich ELISA
Sandwich ELISA Kit
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Nectin-1/PVRL1.
Sandwich ELISA
5818
Rabbit
Storage is content dependent.
23.45-1500 pg/mL
HRP
Antibody Pairs
Rat
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
NECTIN1
RUO
Nectin-1/PVRL1
Videos
SDS
Documents

Documents

Product Certifications