missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nectin-2/CD112 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
£341.00 - £834.00
Specifications
Assay Range | 15.625 pg/mL |
---|---|
Assay Sensitivity | 15.625-1000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18276226
|
Novus Biologicals™
NBP2-79431-15PLATES |
15 Plates |
£834.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18677151
|
Novus Biologicals™
NBP2-79431-5Plates |
5 Plates |
£341.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112.Specifications
15.625 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
NECTIN2 | |
Storage is content dependent. |
15.625-1000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112. | |
Sandwich ELISA | |
5819 | |
Rabbit | |
RUO | |
Nectin-2/CD112 |