missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Needle-Valve Stopcock and Burette Lateral Stopcock
Stopcock with interchangeable PTFE valve spindle, a cap, and a retention device.
Brand: BRAND™ 82121
41.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Valve housing with precision Borosilicate Glass 3.3 burette tip
- Interchangeable PTFE valve spindle, PP cap, and retention device
- Valve opening 0 to 2.5 mm

Specifications
Stopcock | |
0 to 2.5 mm | |
25 mL Burette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Needle-Valve Stopcock and Burette Lateral Stopcock > For Use With: 25 mL Burette
Spot an opportunity for improvement?Share a Content Correction