missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neocuproine Hydrochloride Monohydrate, 99%, Thermo Scientific Chemicals
£56.28 - £617.07
Chemical Identifiers
CAS | 303136-82-5, 484-11-7 |
---|---|
Molecular Formula | C14H12N2 |
Molecular Weight (g/mol) | 208.26 |
MDL Number | MFCD00150062 |
InChI Key | IYRGXJIJGHOCFS-UHFFFAOYSA-N |
Synonym | 2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 % |
PubChem CID | 2723838 |
IUPAC Name | 2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride |
SMILES | CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10255720
|
Thermo Scientific Acros
153310050 |
5 g |
£56.28
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10061840
|
Thermo Scientific Acros
153310250 |
25 g |
£190.95
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10105833
|
Thermo Scientific Acros
153311000 |
100 g |
£617.07
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
303136-82-5, 484-11-7 | |
208.26 | |
IYRGXJIJGHOCFS-UHFFFAOYSA-N | |
2723838 | |
CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 |
C14H12N2 | |
MFCD00150062 | |
2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 % | |
2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride |
Specifications
303136-82-5, 484-11-7 | |
100.0 | |
Yellow | |
99% | |
C14H12N2 | |
2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 % | |
CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 | |
208.26 | |
262.75 | |
Fine Crystalline Powder |
98.5 | |
250°C | |
Authentic | |
Glass Bottle | |
MFCD00150062 | |
IYRGXJIJGHOCFS-UHFFFAOYSA-N | |
2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride | |
2723838 | |
99% | |
Neocuproine hydrochloride monohydrate |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title