missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean FT

Neodisher LaboClean FT - Alkaline detergent liquid for use in special washing machines. For automated cleaning of laboratory glassware in microbiology, virology, nuclear medicine, the food industries, as well as bottles and equipment made of glass in pharmacies.

Brand:  Dr.Weigert™ 410030

Additional Details : CAS Number : 7681-52-9 Weight : 11.96000kg Transport : UN number : 1719 Chem class : 8 Pack group : II


Product Code. 11703733

  • £105.00 / 9.16 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Requires : Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Description

Description

  • Alkaline detergent, liquid concentrate
  • Neodisher LaboClean FT is used for the removal of nutrient mediums, blood, proteins, radioactive contaminants and residues of tissue cultures
  • Free of surfactants and other surface-active substances
  • Oxidative action from active chlorine
  • Suitable for utensils of glass, ceramics, quartz and plastics
  • Dosage of 3-4 m/L
  • Not suitable for non-ferrous metals and light metal alloys

Order Info

Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Certifications

Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Compliance

Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Services

Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Alkaline; detergent; proteins; contaminants, radioactive; chlorine

Specifications

Specifications

Neodisher FT
11.8 to 12.2
Chlorine-based bleaching agents, phosphates
Canister
310 x 230 x 180 mm
Removes Residues of Culture Media, Blood, Proteins, Radioactive Contamination, Residues of Cell and Tissue Cultures as Well as Other Organic Residues
Liquid
Non-sterile
2 to 4 mL/L
1.3 g/cm3 (20°C)
Filtered
12 kg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • neodisher LaboClean FT

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,sodium hypochlorite

SDS
Documents

Documents

Certificates
Special Offers

Special Offers