missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LM2

Neodisher LM 2 - Midly alkaline cleaning agent liquid, manual cleaning of laboratory glassware in immersion baths in Medical, dental practices, industrial and Biological laboratories. Can be used in immersion and ultra sonic baths.

Brand:  Dr.Weigert™ 440330

Additional Details : Weight : 0.80000kg


Product Code. 11833242

  • £146.00 / Each
In Stock
to see stock.

Description

Description

  • Mildly alkaline liquid cleaning agent
  • Neodisher LM2 is used for manual cleaning laboratory glassware, surgical instruments, utensils and cages for experimental animals
  • Can be used in immersion and ultra sonic bath
  • Offers high protection for laboratory glass, stainless steel, instrument steel and plastics
  • Aluminum and light metals should be tested first
  • Effective against encrusted, protein blood, inorganic residues and cement dental
  • Not create foam
  • Suited for use in special washers for e.g. viscosimeters or pipettes
  • Can be used with all water hardness levels
  • Suggested dose is 5-30 mL/L
  • Specifications

    Specifications

    Neodisher LM 2
    10.6 to 11.3
    Amphoteric surfactants; anionic surfactants, phosphates
    1.2 g/cm3
    Filtered
    Liquid
    Non-sterile
    5 to 30 mL/L
    310 x 230 x 180 mm
    10 L
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • neodisher LM 2

    Signal Word
    • Warning

    Hazard Category
    • Skin corrosion/irritation Category 2
    • Serious eye damage/eye irritation Category 2

    Hazard Statement
    • H315-Causes skin irritation.
    • H319-Causes serious eye irritation.
    • EUH208-Contains . May produce an allergic reaction.

    Precautionary Statement
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P302+P352-IF ON SKIN: Wash with plenty of water/.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P337+P313-If eye irritation persists: Get medical advice/attention.

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers