Learn More
Dr.Weigert™ Neodisher PolyKlar
Neodisher PolyKlar - Rinse aid liquid. Free of surfactants. Neutralizes alkaline residues during automated reprocessing. Designed for rinsing of animal cages.
Brand:  Dr.Weigert™ 401930
Additional Details : Weight : 10.00000kg
Product Code. 10788985
  • £138.00 / Each
In Stock
to see stock.
Description
Description
  • Rinse aid Without surfactant
  • neodisher PolyKlar is used in specially designed washers for animal cages and accessories
  • Dosage of between 0.5-2 mL/L
  • Acidic formulation
  • Excellent material compatibility eg polysulfones, polycarbonates, polyphenylsulfones and polyetherimides
  • Neutralizes alkaline residues
Specifications
Specifications
Neodisher Polyklar
3.6 to 3.2
Polycarboxylates, chloromethylisothiazolinones, methylisothiazolinones
1.0 g/cm3 (20°C)
Filtered
Liquid
Non-sterile
0.5 to 2 mL/L
310 x 230 x 180 mm
10 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • neodisher PolyKlar
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : disodium metasilicate pentahydrate, sodium carbonate, troclosene sodium