missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher Z

Neodisher Z - Acidic neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent during automated reprocessing. Free of surfactants and phosphates.

Brand:  Dr.Weigert™ 420235


 View more versions of this product

Product Code. 11748393

  • £112.00 / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Based on citric acid
  • neodisher Z is used for neutralizing alkaline residues in washer disinfection
  • Without phosphates, phosphoric acid, nitrogens compounds and surfactants
  • Dosage of between 2-4 mL/L
  • Gentle on materials
  • Laboratory glassware in medical, industrial and water laboratories
Specifications

Specifications

Neodisher Z
3 to 2.6
Citric acid, 2-octyl-2 H-isothiazol-3-one
Canister (High Form)
5 L
Liquid
Non-sterile
1 to 5 mL/L
250 x 190 x 140 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • neodisher Z

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : citric acid, anhydrous

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.