missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Description
Neodymium applications include lasers, glass coloring and tinting, dielectrics and, most importantly, as the fundamental basis for neodymium-iron-boron (Nd2Fe14B) permanent magnets. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
UN Number | UN3178 |
Quantity | 25 x 25 mm |
Solubility Information | Reacts with water. |
Percent Purity | 99.9% |
Odor | Odorless |
Packaging | Mylar under Argon |
Weight | ≈0.43g/25x25mm |
Form | Foil |
Assay | (REO) |
Chemical Name or Material | Neodymium foil, 0.1mm (0.004 in.) thick |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'