missing translation for 'onlineSavingsMsg'
Learn More

Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11400390
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm

Neodymium applications include lasers, glass coloring and tinting, dielectrics and, most importantly, as the fundamental basis for neodymium-iron-boron (Nd2Fe14B) permanent magnets. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

UN Number UN3178
Quantity 50 x 50 mm
Solubility Information Reacts with water.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.43g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Neodymium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'