missing translation for 'onlineSavingsMsg'
Learn More
Neodymium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11395759

Neodymium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11395759
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11395759

Brand: Thermo Scientific Alfa Aesar 010248.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium foil is used in alloys to produce permanent magnets and flints. It is also used as an additive to glasses which are used in solid state lasers and light amplifiers. It is also employed in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films. The neodymium-based magnet is used extensively in the automotive industry with many applications including starter motors, brake systems, seat adjusters and car stereo speakers. It is also used in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Quantity 50 x 50 mm
Solubility Information Soluble in dilute acids. Decomposes in water.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈4.4g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Neodymium foil, 1.0mm (0.04 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS07I036 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog Number010248.FF,010248.FI
Lot NumberE14W037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010248.FF,010248.FI
Lot NumberU06H022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010248.FF,010248.FI
Lot NumberF06X003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010248.FF,010248.FI
Lot NumberW22B002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010248.FF,010248.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.