missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11382278

Neodymium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11382278
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11382278

Brand: Thermo Scientific Alfa Aesar 013184.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14589-38-9
Molecular Formula C15H21NdO6
Molecular Weight (g/mol) 441.57
MDL Number MFCD00013498
InChI Key AZTDRGMPTCCVFW-UHFFFAOYSA-N
Synonym Neodymium acetylacetonate
PubChem CID 122177161
SMILES [Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 150°C to 152°C
Physical Form Powder
Quantity 2 g
IUPAC Name neodymium(3+) tris(2,4-dioxopentan-3-ide)
Formula Weight 441.57
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Neodymium(III) 2,4-pentanedionate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD18X043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013184.04,013184.09
Lot NumberI19K04 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013184.04,013184.09
Lot NumberF16W017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013184.04,013184.09
Lot NumberH21Y046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013184.04,013184.09
Lot NumberZ22D019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013184.04,013184.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.