missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11445370
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
334869-71-5
C6H9NdO6
321.37
MFCD00013040
KETUDCKOKOGBJB-UHFFFAOYSA-K
neodymium(3+) triacetate
[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
334869-71-5
C6H9NdO6
321.37
MFCD00013040
KETUDCKOKOGBJB-UHFFFAOYSA-K
neodymium(3+) triacetate
[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 321.38 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium (III) acetate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'