missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals Product Code.: 10563041

Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Product Code. 10563041
5 g, Glass bottle
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10563041

Brand: Thermo Scientific Acros 315070050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 784°C
CAS 10024-93-8
Color White
CAS Min % 99.85
CAS Max % 100.0
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.90%
Molecular Formula Cl3Nd
Linear Formula Cl3Nd
MDL Number MFCD00011132
Merck Index 10, 6295
Solubility Information Solubility in water: 96.7g/100mL (13°C)
Molecular Weight (g/mol) 250.59
Formula Weight 250.59
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Neodymium(III) chloride, 99.9%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398022 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number315070050
Lot NumberA0414046 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number315070050
Lot NumberA0434907 Certificate TypeCertificate of Analysis Date02/10/2021 Catalog Number315070050
Lot NumberA0423928 Certificate TypeCertificate of Analysis Date02/11/2020 Catalog Number315070050
Lot NumberA0423443 Certificate TypeCertificate of Analysis Date15/10/2020 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.