missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11306318

Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11306318
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11306318

Brand: Thermo Scientific Alfa Aesar 011251.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-78-0
MDL Number MFCD00011132

Specifications

Melting Point 124°C
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.9% (REO)
Molecular Formula NdCl3·H2O
Solubility Information Soluble in water.
Formula Weight 250.60 (Anhydrous)
Odor Odorless
Appearance Crystalline aggregrates
Chemical Name or Material Neodymium(III) chloride hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20J044 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number011251.18,011251,011251.30
Lot NumberH16X027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011251.18,011251.30
Lot NumberU16H024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011251.18,011251.30
Lot NumberM13A015 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011251.18,011251.30
Lot NumberB15X026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011251.18,011251.30
compliance-icons
Product Identifier
  • Neodymium(III) chloride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Neodymium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.