missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11366918

Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11366918
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11366918

Brand: Thermo Scientific Alfa Aesar 013661.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium(III) fluoride, anhydrous is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-42-7
Molecular Formula F3Nd
Molecular Weight (g/mol) 201.24
MDL Number MFCD00011135
InChI Key XRADHEAKQRNYQQ-UHFFFAOYSA-K
IUPAC Name neodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Nd+3]

Specifications

Melting Point 1410°C
Color Purple
Physical Form Crystalline Lump
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 201.24
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 6.5 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) fluoride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX22K019 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number013661.14,013661.22
Lot NumberU09C022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013661.14,013661.22
Lot NumberR01A036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013661.14,013661.22
Lot NumberE05Q47A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013661.14,013661.22
Lot NumberQ06B029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013661.14,013661.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.