missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neodymium(III) hexafluoro-2,4-pentanedionate dihydrate, Thermo Scientific™
£30.00 - £75.10
Chemical Identifiers
CAS | 47814-18-6 |
---|---|
Molecular Formula | C15H7F18NdO8 |
Molecular Weight (g/mol) | 801.43 |
MDL Number | MFCD00167152 |
InChI Key | NBCIEIIVJWSHPT-UHFFFAOYSA-N |
Synonym | neodymium lll hexafluoroacetylacetonate dihydrate, neodymium hexafluoropentanedionate dihydrate, neodymium iii hexafluoro 2,4-pentanedionate, neodymium iii hexafluoroacetylacetonate dihydrate, neodymium iii hexafluoro-2,4-pentanedionate dihydrate, neodymium iii hexafluoroacetylacetonate dihydrate, ?nd cf3cochcocf3 3.2h2o, neodymium,tris 1,1,1,5,5,5-hexafluoro-2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy neodymio oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one dihydrate |
IUPAC Name | neodymium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide) dihydrate |
SMILES | O.O.[Nd+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
47814-18-6 | |
801.43 | |
NBCIEIIVJWSHPT-UHFFFAOYSA-N | |
neodymium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide) dihydrate |
C15H7F18NdO8 | |
MFCD00167152 | |
neodymium lll hexafluoroacetylacetonate dihydrate, neodymium hexafluoropentanedionate dihydrate, neodymium iii hexafluoro 2,4-pentanedionate, neodymium iii hexafluoroacetylacetonate dihydrate, neodymium iii hexafluoro-2,4-pentanedionate dihydrate, neodymium iii hexafluoroacetylacetonate dihydrate, ?nd cf3cochcocf3 3.2h2o, neodymium,tris 1,1,1,5,5,5-hexafluoro-2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy neodymio oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one dihydrate | |
O.O.[Nd+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F |
Specifications
47814-18-6 | |
C15H7F18NdO8 | |
neodymium lll hexafluoroacetylacetonate dihydrate, neodymium hexafluoropentanedionate dihydrate, neodymium iii hexafluoro 2,4-pentanedionate, neodymium iii hexafluoroacetylacetonate dihydrate, neodymium iii hexafluoro-2,4-pentanedionate dihydrate, neodymium iii hexafluoroacetylacetonate dihydrate, ?nd cf3cochcocf3 3.2h2o, neodymium,tris 1,1,1,5,5,5-hexafluoro-2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy neodymio oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one dihydrate | |
O.O.[Nd+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F | |
801.43 | |
Neodymium(III) hexafluoro-2,4-pentanedionate dihydrate |
Crystalline | |
MFCD00167152 | |
NBCIEIIVJWSHPT-UHFFFAOYSA-N | |
neodymium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide) dihydrate | |
801.42 (765.39 Anhydrous) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures