missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11316308

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11316308
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11316308

Brand: Thermo Scientific Alfa Aesar 011245.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-96-8
Molecular Formula N3NdO9
Molecular Weight (g/mol) 330.25
MDL Number MFCD00149798
InChI Key CFYGEIAZMVFFDE-UHFFFAOYSA-N
Synonym neodymium iii nitrate hydrate, neodymium nitrate hydrate, acmc-20ak53, 3no3.nd.h2o, neodymium nitrate-water 1/3/1, neodymium 3+ ion hydrate trinitrate, neodymium 3+ hydrate trinitrate, neodymium iii nitrate hydrate, reacton, nitric acid,neodymium 3+ salt, hydrate 8ci,9ciShow MoreShow Less
PubChem CID 71311426
SMILES [Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∽40°C
Color Blue
Physical Form Wet Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
IUPAC Name neodymium(3+);trinitrate;hydrate
Formula Weight 330.25 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Violet crystals
Chemical Name or Material Neodymium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ07G005 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number011245.06,011245.14
Lot Number10251662 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011245.06,011245.14
Lot NumberB27U011 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number011245.06,011245.14
Lot Number61200628 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number011245.06,011245.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.