missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neodymium(III)-oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals
£45.83 - £104.52
Chemical Identifiers
CAS | 1313-97-9 |
---|---|
Molecular Formula | Nd2O3 |
Molecular Weight (g/mol) | 336.48 |
MDL Number | MFCD00011134 |
InChI Key | PLDDOISOJJCEMH-UHFFFAOYSA-N |
IUPAC Name | dineodymium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Nd+3].[Nd+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10686802
|
Thermo Scientific Acros
211580250 |
25 g | Glass bottle |
£45.83
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606982
|
Thermo Scientific Acros
211581000 |
100 g | Glass bottle |
£104.52
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
1313-97-9 | |
336.48 | |
PLDDOISOJJCEMH-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Nd+3].[Nd+3] |
Nd2O3 | |
MFCD00011134 | |
dineodymium(3+) trioxidandiide |
Specifications
2270.0°C | |
Blue | |
100.0 | |
99.90% | |
ND2O3 | |
14, 6450 | |
PLDDOISOJJCEMH-UHFFFAOYSA-N | |
dineodymium(3+) trioxidandiide | |
336.47 | |
Trace Metal Basis | |
7.2400g/mL |
1313-97-9 | |
99.85 | |
Powder | |
Nd2O3 | |
MFCD00011134 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in acids | |
[O--].[O--].[O--].[Nd+3].[Nd+3] | |
336.48 | |
99.9% | |
7.24 | |
Neodymium(III)-oxide, 99.9% |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 215-214-1
missing translation for 'rtecsNumber' : QP0185000
missing translation for 'tsca' : TSCA
Product Identifier
- Neodymium(III)-oxide
RUO – Research Use Only