missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals Product Code.: 11430291

Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Product Code. 11430291
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11430291

Brand: Thermo Scientific Alfa Aesar 040568.NB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17522-69-9
Molecular Formula Cl3NdO12
Molecular Weight (g/mol) 442.58
MDL Number MFCD00198195
InChI Key HBKMAYJLXKBOER-UHFFFAOYSA-K
Synonym neodymium 3+ perchlorate, neodymium perchlorate, neodymium 3+ triperchlorate, neodymium 3+ ion triperchlorate ion, neodymium iii perchlorate aqueous solution
PubChem CID 13783607
IUPAC Name neodymium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Nd+3]

Specifications

Physical Form Liquid
Quantity 5 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 550.68 (442.59 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Neodymium(III) perchlorate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ29I015 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number040568.NB,040568.ND
Lot NumberW24F041 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040568.NB,040568.ND
Lot NumberT30F069 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040568.NB,040568.ND
Lot NumberJ16R052 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040568.NB,040568.ND
Lot NumberZ05C084 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040568.NB,040568.ND

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.