missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 25 g |
UN Number | UN3211 |
Solubility Information | Insoluble in water. |
Formula Weight | 550.68 (442.59 Anhydrous) |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Chemical Name or Material | Neodymium(III) perchlorate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'