Learn More
Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Description
Neodymium(III) trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes and as a catalyst for dehydration of fructose, Diels-Alder reactions of cyclopentadiene and alkyl acrylates, α-amination reactions of pyrazolones, polymerization reactions of cyclosiloxanes, direct polycondensation of dicarboxylic acids and diols, regioselective reductive ring opening of benzylidene acetals, Paal-Knorr cyclocondensation and nitration of benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | Soluble in water. |
Formula Weight | 591.44 |
Percent Purity | 98% |
Density | 1.7 g/mL |
Chemical Name or Material | Neodymium (III) trifluoromethanesulfonate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T19J036 | Certificate of Analysis | 25/07/2023 | 043393.18,043393.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.