missing translation for 'onlineSavingsMsg'
Learn More

Neodymium ingot, 99.1% (metals basis), Thermo Scientific Chemicals

Product Code. 11300699
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray-Silver
Quantity 25 g
Percent Purity 99.1%
Form Ingot
Assay (metals basis)
Chemical Name or Material Neodymium ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'