missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Neomycin Sulfate
Acts as an inhibitor of protein biosynthesis. Fisher BioReagents™ Neomycin Sulfate is a white powder used in tissue culture studies.
Brand: Fisher BioReagents™ 12787149
This item is not returnable.
View return policy
Description
- Purity 90-95% Neomycin B
- Potency ≥630μg neomycin/mg
- FTIR Conforms to standard
- pH (3.3% aqueous solution) 5-7.5
- Loss on drying ≤8%
Specifications
Store in a cool, dry, dark place | |
Antibiotic | |
5 to 7.5 | |
5 g |
Potency: ≥630 μg/mg | |
Amber bottle | |
90-95% Neomycin B |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Neomycin Sulfate
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Neomycin sulfate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For research and laboratory use only. NJot for drug use.
Spot an opportunity for improvement?Share a Content Correction