Learn More
Invitrogen™ Neon™ Transfection System 10 μL Kit
Neon™ Transfection System 10 μL Kit
Brand: Invitrogen™ MPK1096
1433.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Includes: 3 x 1mL Resuspension Buffer R, 3 x 1mL Resuspension Buffer T, 2 x 150mL Electrolytic Buffer E, 96 x 10μL Neon tips, 20 Neon electroporation tubes
Description
- Animal origin-free, serum-free and designed specifically for use with the Neon Transfection System
- Used for transfection volumes of 10μL, containing 5 x 104-2 x 105 adherent cells or 1 x 106-5 x 105 suspension cells
- Cells that have been transfected using included 10μL Neon Tips are ready to be washed and plated into multiwell culture dish
Adenoviral shRNA, Cell Culture, Lentiviral shRNA & miRNA, Plasmid Transfection, RNAi, RNAi Transfection, RNAi, Epigenetics & Non-Coding RNA Research, Retroviral RNAi, Stem Cell & Primary Cell Transfections, Synthetic siRNA Transfection, Transfection, Vector-Based RNAi, siRNA
Order Info
Shipping Condition: Room Temperature
Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum | |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA | |
10 x 10^4 - 2 x 10^5 cells/mL (Adherent Cells), 1 x 10^6 - 5 x 10^5 cells/mL | |
Transfection | |
Neon™ | |
96 x 2 Reactions | |
Room Temperature | |
Neon™ Transfection System |
• 3 x 1 mL Resuspension Buffer R • 3 x 1 mL Resuspension Buffer T • 2 x 150 mL Electrolytic Buffer E • 96 x 10 μl Neon™ tips • 20 Neon™ electroporation tubes Store the buffer box 4°C and the tips and tubes box at room temperature. |
|
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Electroporation | |
Neon™ Transfection System | |
96 x 2 | |
Transfection System | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type | |
10μl, 10 μl |
For Research Use Only. Not for use in diagnostic procedures.