missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ Transfection System 10 μL Kit

Product Code. 10698888
Click to view available options
No. of Reactions:
25 x 2
96 x 2
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10698888

Brand: Invitrogen™ MPK1025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Neon™ Transfection System 10 μL Kit

The Neon™ Transfection System 10 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 10 μL, containing 5 × 104–2 × 105 adherent cells or 1 × 105–5 × 105 suspension cells. Cells that have been transfected using the included 10μL Neon™ Tips are ready to be washed and plated into a multi-well culture dish.
TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Content And Storage The Neon™ Transfection System 10μL Kit comes with 1 mL resuspension buffer R, 1 mL resuspension buffer T, and 75 mL E electrolytic buffer in one box and 25 reaction delivery tips and 5 electroporation tubes in a separate box. Refer to the Neon™ Transfection System users manual for protocol instructions. Store the buffer box 4°C and the tips & tubes box at room temperature.
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA
Transfection Technique Electroporation
For Use With (Equipment) Neon™ Transfection System
For Use With (Application) Transfection
No. of Reactions 25 x 2
Product Line Neon
Product Type Transfection System
Quantity 25 x 2 Reactions
Serum Compatible Yes
Shipping Condition Room Temperature
Starting Material Amount 2x10^4 - 6 x10^8 cells; Varies by cell type
System Type Neon™ Transfection System
Volume (Metric) Sample 10 μl
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.