missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neon™ Transfection System 100 μL Kit
Neon™ Transfection System 100 μL Kit
£622.00 - £1764.00
Specifications
Classification | Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum |
---|---|
Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Plasmid DNA, Synthetic siRNA |
Transfection Technique | Electroporation |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10114334
|
Invitrogen™
MPK10096 |
96 x 2 |
£1764.00
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704289
|
Invitrogen™
MPK10025 |
50 Reactions |
£622.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
The Neon™ Transfection System 100 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 100 μL, containing 5 × 105–2 × 106 adherent cells or 1 × 106–5 × 106 suspension cells. Cells that have been transfected using the included 100μL Neon™ Tips are then ready to be washed and plated into a 6-well, 60 mm, or 10 cm culture dish, depending on your protocol.Spezifikation
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Electroporation | |
Transfection | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type | |
100μl |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA | |
Neon™ Transfection System | |
Neon™ | |
Room Temperature | |
Neon™ Transfection System |
For Research Use Only. Not for use in diagnostic procedures.