Learn More
Invitrogen™ Neon™ Transfection System Pipette Station
Neon™ Transfection System Pipette Station
Brand: Invitrogen™ MPS100
Code : 50
Additional Details : Weight : 0.44000kg
Description
The Neon™ Transfection System Pipette Station is designed specifically for use with the Neon™ Transfection System device (MPK5000). Each device (MPK5000) already comes with a pipette station. The pipette station is also sold separately for those labs needing and additional pipette station or replacement for their existing pipette station.
For more information on the product, please click here
Specifications
The Neon™ Transfection System Pipette station comes with 1 x pipette station. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions. | |
Neon | |
Neon™ Transfection System | |
Room Temperature | |
Neon™ Transfection System |
Transfection Grade Nucleic Acid Purification Kits, siRNA Vectors, DNA Vectors | |
Electroporation Pipette Tip | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type |
For Research Use Only. Not for use in diagnostic procedures.