Learn More
Invitrogen™ Neon™ Transfection System Pipette
Neon™ Transfection System Pipette
Brand: Invitrogen™ MPP100
574.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10716kg
Description
The Neon™ Transfection System Pipette is designed specifically for use with the Neon™ Transfection System device (MPK5000). Each device (MPK5000) already comes with a pipette. The pipette is also sold separately for those labs needing and additional pipette or replacement for their existing pipette.
For more information on the product, please click here
Specifications
The Neon™ Transfection System Pipette comes with 1 x pipette. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions. | |
Neon™ | |
Neon™ Transfection System | |
Room Temperature | |
Neon™ Transfection System |
Transfection Grade Nucleic Acid Purification Kits, siRNA Vectors, DNA Vectors | |
Electroporation Pipette Tip | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type |
For Research Use Only. Not for use in diagnostic procedures.