missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Neoprene Gloves

Product Code. 11747385 Shop All SP Scienceware Products
Small
Click to view available options
Size:
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

One piece, bonded neoprene protection against liquid or gas penetration

  • Use for glove boxes with 8 in. glove ports
  • Requires clamping rings to hold gloves in place

Specifications

Description Sleeved Gloves
Color Black
Material Bonded Neoprene
Product Type Gloves
Size Small
For Use With (Equipment) Glove Boxes with 8 in. Ports
Quantity 2 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Neoprene Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.