Learn More
Fisherbrand™ Solid Neoprene Stoppers
Offer resistance to heat, ozone and petroleum based oils.
£9.25 - £36.00
Product Code | Brand | Diameter (Metric) Bottom | Height (Metric) | Stopper Number | Diameter (Metric) Top | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Height (Metric) | Stopper Number | Diameter (Metric) Top | Price | Quantity & Availability | ||
11587103
|
Fisherbrand™
|
37 mm | 38.5mm | 37 | 42.5 mm |
£29.85
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11577093
|
Fisherbrand™
|
15 mm | 24mm | 15 | 18 mm |
£17.00
Pack of 20 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11557103
|
Fisherbrand™
|
31 mm | 35mm | 31 | 36 mm |
£21.65
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11577103
|
Fisherbrand™
|
35 mm | 38.5mm | 35 | 40.5 mm |
£14.60
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11507113
|
Fisherbrand™
|
41 mm | 42mm | 41 | 47 mm |
£9.70
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11597083
|
Fisherbrand™
|
5 mm | 16mm | 5 | 7 mm |
£9.25
Pack of 20 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11527113
|
Fisherbrand™
|
45 mm | 42mm | 45 | 51 mm |
£20.35
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11527093
|
Fisherbrand™
|
8 mm | 20mm | 8 | 10.5 mm |
£11.90
Pack of 20 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11507103
|
Fisherbrand™
|
21 mm | 28mm | 21 | 24.5 mm |
£27.50
Pack of 10 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11517103
|
Fisherbrand™
|
23 mm | 28mm | 23 | 26.5 mm |
£32.50
Pack of 10 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11547103
|
Fisherbrand™
|
29 mm | 32mm | 29 | 33 mm |
£36.00
Pack of 10 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
Description
- Better resistance to heat, ozone and petroleum based oils than red rubber
- Less suitable for low temperature use
- Operational temperature range -30°C to 100°C
Specifications
Gray | |
Neoprene | |
Solid Stopper |