missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neosel

Neosel - Regeneration Salt is specially formulated to obtain a brine. For professional and domestic dishwashers.

Brand:  Dr.Weigert™ 600012

Product Code. 11758393

  • £18.20 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Regeneration Salt
  • Neosel is used for dishwasher and washer disinfection with integrated water softener
  • Very high purity
  • Mechanically compressed
  • Granulometry 1 to 3 mm
  • Recrystallization sodium chloride obtained by evaporation under vacuum
Specifications

Specifications

Regeneration Salt
Non-sterile
Bucket
150 x 230 mm
Crystalline
Sodium chloride
1.100 - 1.300 kg/m3
5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.