missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Neox™ Black Neoprene Immersion Gloves

Protects against a wide range of oils, acids, caustics, alcohols and many solvents. Comfortable anatomical glove shape follows the contours of the hand at rest. Improves dexterity and reduces hand fatigue.

Brand:  Ansell Edmont™ 9928100

Product Code. 12629801

  • £448.00 / 12 pairs
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Top quality neoprene coating.
  • Good resistance to all mechanical hazards.
  • Special Hi-Lo insulated gauntlet option with quilted cotton liner, for use in both hot and cold temperatures.
  • Permits intermittent handling in hot liquids up to 180°C and handling in cold temperatures down to -25°C
  • Comfortable anatomical glove shape follows the contours of the hand at rest.
  • Improves dexterity and reduces hand fatigue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neoprene
Neoprene
Immersion Gloves
180°C
Cotton Jersey
1 Pair
09-928
Standard CATEGORY III, EN 374, EN 388 : 3121, ANSI, FDA 21 CFR 177.2600
Neoprene
09-928
Gauntlet
Fully coated-palm
Excellent broad-spectrum chemicals
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.