Learn More
Nepafenac-d5, MedChemExpress
MedChemExpress Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.
Brand: Medchem Express HY-17357S
This item is not returnable.
View return policy
Description
Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.Specifications
Nepafenac-d5 | |
5 mg | |
Research | |
AHR-9434-d5 AL-6515-d5 | |
O=C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=CC=CC(CC(N)=O)=C2N | |
259.31 | |
Research |
1246814-53-8 | |
Yellow | |
C15H9D5N2O2 | |
DMSO : ≥ 100 mg/mL (385.64 mM) | |
259.31 | |
98.0% | |
Solid |
Research purposes only