missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Acetyl-Nalpha-Boc-L-lysine, 98%, Thermo Scientific™

£16.07 - £144.00

Chemical Identifiers

CAS 6404-26-8
Molecular Formula C13H24N2O5
Molecular Weight (g/mol) 288.34
MDL Number MFCD00057791
InChI Key IOKOUUAPSRCSNT-UEQNJFAPNA-N
Synonym boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
PubChem CID 7016049
IUPAC Name (2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15408878
View Documents
Thermo Scientific Alfa Aesar
H62347.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15498868
View Documents
Thermo Scientific Alfa Aesar
H62347.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6404-26-8
288.34
IOKOUUAPSRCSNT-UEQNJFAPNA-N
7016049
CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
C13H24N2O5
MFCD00057791
boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
(2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid

Specifications

6404-26-8
MFCD00057791
IOKOUUAPSRCSNT-UEQNJFAPNA-N
(2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
7016049
98%
C13H24N2O5
boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
288.34
288.34
Nepsilon-Acetyl-Nalpha-Boc-L-lysine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications