missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Benzyloxycarbonyl-D-lysine, 95%, Thermo Scientific Chemicals

Product Code. 15419288
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34404-32-5
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00063120
InChI Key CKGCFBNYQJDIGS-STGVRZAANA-N
Synonym h-d-lys z-oh, n6-phenylmethoxy carbonyl-r-lysine, r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 2r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, n-e-z-d-lysine, ambotzhaa6320, nepsilon-cbz-d-lysine, d-lys z-oh, nepsilon-carbobenzoxy-d-lysine, n6-benzyloxycarbonyl-d-lysine
PubChem CID 1551330
IUPAC Name (2R)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Formula Weight 280.32
Percent Purity 95%
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-D-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'