missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride, 98%, Thermo Scientific™
£21.60 - £76.30
Chemical Identifiers
CAS | 6366-70-7 |
---|---|
Molecular Formula | C21H27ClN2O4 |
Molecular Weight (g/mol) | 406.91 |
MDL Number | MFCD00038981 |
InChI Key | NWSFLBWTZZJOAF-FYZYNONXSA-N |
Synonym | h-lys z-obzl.hcl, n6-cbz-l-lysine benzyl ester hydrochloride, h-lys z-obzl hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-obzl hcl, h-lys z-obzl inverted exclamation mark currencyhcl, lys z-obzl.hcl, benzyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hcl, lys z-obzl hcl |
PubChem CID | 11750132 |
IUPAC Name | benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride |
SMILES | Cl.N[C@@H](CCCCNOC(=O)CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468466
|
Thermo Scientific Alfa Aesar
H63714.03 |
1 g |
£21.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478466
|
Thermo Scientific Alfa Aesar
H63714.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6366-70-7 | |
406.91 | |
NWSFLBWTZZJOAF-FYZYNONXSA-N | |
11750132 | |
Cl.N[C@@H](CCCCNOC(=O)CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
C21H27ClN2O4 | |
MFCD00038981 | |
h-lys z-obzl.hcl, n6-cbz-l-lysine benzyl ester hydrochloride, h-lys z-obzl hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-obzl hcl, h-lys z-obzl inverted exclamation mark currencyhcl, lys z-obzl.hcl, benzyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hcl, lys z-obzl hcl | |
benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride |
Specifications
6366-70-7 | |
C21H27ClN2O4 | |
3580519 | |
NWSFLBWTZZJOAF-FYZYNONXSA-N | |
benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride | |
11750132 | |
98% | |
Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride |
White | |
MFCD00038981 | |
h-lys z-obzl.hcl, n6-cbz-l-lysine benzyl ester hydrochloride, h-lys z-obzl hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-obzl hcl, h-lys z-obzl inverted exclamation mark currencyhcl, lys z-obzl.hcl, benzyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hcl, lys z-obzl hcl | |
Cl.N[C@@H](CCCCNOC(=O)CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 | |
406.91 | |
406.9 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 228-859-9
missing translation for 'rtecsNumber' : OL5685000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold