missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nessler Cylinder

Manufactured from soda lime glass. Fisherbrand™ Nessler Cylinders are ideal for colorimetric analysis.

Brand:  Fisherbrand™ FB68961

 View more versions of this product

Product Code. 11932168

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For comparison of color and turbidity between liquids or solutions
  • Fixed values of 50mL or 100mL with 50mL graduation
  • Optically plane bottom provides maximum clarity for visual comparisons
  • Supplied in matched pairs to ensure uniformity in product for accuracy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.8 mL
Flat
100 mL
Nessler Tube
Glass
34 mm
180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers