Learn More
Ohaus™ Nester Clamp
OHAUS™ Nester Clamp designed to securely grip and position laboratory glassware.
Brand: Ohaus™ 30392318
Additional Details : Weight : 0.50000kg
Description
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Single Adjust
Specifications
Stainless Steel | |
10 mm | |
5.980 in. | |
152 mm | |
Nester |
Clamp, Specialty, Nester, CLS-NESTXS | |
10.32 in. | |
262.00 mm | |
50 to 102 mm |