Learn More
Emma Safety Footwear Nestor Safety Shoes
Oiled nubuck leather is soft to the touch yet repels water like a duck. The ideal in-and-out shoe is perfect for visiting a construction site.
Brand: Emma Safety Footwear MM76254638
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black, Brown | |
Hydro-Tec™ Sanitized Silver | |
38 |
Laces | |
Nestor | |
Oiled Nubuck | |
Steel |