missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
12435680
|
Cytiva
28-9663-81 |
For 50 mm HiScale Columns I.D. |
£79.00
Pack of 5 |
In Stock
Log in to see stock. |
||||||
11663224
|
Cytiva
28-9663-80 |
For 26 mm HiScale Columns I.D. |
£63.00
Each |
In Stock
Log in to see stock. |
||||||
Description
Specifications
Net Ring | |
5 Pack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Net Ring