missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neurobasal™-A Medium, no D-glucose, no sodium pyruvate
GREENER_CHOICE

Gibco™ Neurobasal™-A medium without D-glucose and sodium pyruvate is used to support the growth of post-natal CNS neurons in applications where the presence of D-glucose and sodium pyruvate can cause interference.

Brand:  Gibco™ A2477501

57.50 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 15329741

  • £86.25 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

  • For neural studies that require the absence of insulin, glucose, and sodium pyruvate, such as glucose response or insulin response studies, we recommend Gibco B-27 Supplement Minus Insulin as a supplement to this medium.
  • Neurobasal Medium is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of Neurobasal media and B-27 supplements.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C) and protect from light.
Serum-free
Liquid
Phenol Red
Neurobasal™
Ambient
Neural Cells
Neuronal Cell Culture
Neurobasal Medium
No Glucose, No Sodium Pyruvate
500 mL
Endotoxin, Performance, Sterility, Osmolality, pH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Neurobasal™-A Medium, no D-glucose, no sodium pyruvate > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.