missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neurobasal™-A Medium
GREENER_CHOICE

Product Code. 11540366
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11540366

Brand: Gibco™ 10888022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Neurobasal™-A Medium

Neurobasal™-A Medium is a basal medium formulated to meet the special cell culture requirements of post-natal and adult brain neuronal cells when used with GIBCO™ B-27™ Supplement. Neurobasal™-A Medium can be used to grow neuronal cells from hippocampus, cortex and other regions of the brain. Neurobasal™-A Medium allows for both long and short term maintenance of homogeneous populations of neuronal cells without the need of an astrocyte feeder layer.

Add Your Own Supplements
All Neurobasal™ media contain no L-glutamine, L-glutamic acid, or aspartic acid. Neurobasal™ media must be supplemented with 0.5 mM L-glutamine or GlutaMAX™-I Supplement and either a serum-free supplement (such as B-27™ Supplement or N-2 Supplement), or with serum. For initial plating, you should add 25μM glutamic acid.

Quality Control Testing
All Neurobasal™ media are tested for pH, osmolality, endotoxin, and the absence of bacterial and fungal contamination. The osmolality of Neurobasal™-A was demonstrated to be optimal for postnatal and adult CNS neurons.

Neurobasal™-A Medium contains Phenol Red. If Phenol Red will interfere with your applications, we also provide Neurobasal™-A Medium Minus Phenol Red.

Neurobasal™-A Medium is optimized for post-natal and adult brain neurons. For growing pre-natal and embryonic neurons, we provide Neurobasal™ Medium.

See the entire Neurobasal™ product line.

Learn more about our neural cell culture products.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type Neurobasal Medium
With Additives Phenol Red
Product Line Neurobasal
Quantity 500 mL
Shipping Condition Ambient
Tested For Endotoxin, Performance, Sterility, Osmolality, pH
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Neuronal Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.