missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neurobasal™ Medium, minus phenol red
GREENER_CHOICE

Product Code. 11560426
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560426

Brand: Gibco™ 12348017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Neurobasal™ Medium, minus phenol red

Neurobasal™ Medium Minus Phenol Red is a basal media that meets the special requirements of pre-natal and embryonic neuronal cells when used with GIBCO™ B-27™ Supplement, but without Phenol Red. Neurobasal™ Medium can be used to grow neuronal cells from hippocampus, cortex and other regions of the brain. Neurobasal™ Medium allows for both long and short term maintenance of homogeneous populations of neuronal cells without the need of an astrocyte feeder layer.

Use Neurobasal™ Medium Minus Phenol Red for applications in which the presence of Phenol Red will interfere with your results.

Add Your Own Supplements
All Neurobasal™ media contain no L-glutamine, L-glutamic acid, or aspartic acid. Neurobasal™ media must be supplemented with 0.5 mM L-glutamine or GlutaMAX™-I Supplement and either a serum-free supplement (such as B-27™ Supplement or N-2 Supplement), or with serum. For initial plating, you should add 25μM glutamic acid.

Quality Control Testing
All Neurobasal™ media are tested for pH, osmolality, endotoxin, and the absence of bacterial and fungal contamination.

We also provide Neurobasal™ Medium with Phenol Red.

Neurobasal™ Medium is optimized for pre-natal and embryonic neurons. For growing post-natal and adult brain neurons, we provide Neurobasal™-A Medium

See the entire Neurobasal™ product line.

Learn more about neural cell culture products from Invitrogen at www.invitrogen.com/neuralculture.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type Neurobasal Medium
Without Additives No Phenol Red
Product Line Neurobasal
Quantity 500 mL
Shipping Condition Ambient
Tested For Endotoxin, Performance, Sterility, Osmolality, pH
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Neuronal Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.