missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (McNFn27b) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-154
304.00 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Neurofibromin 1 | |
Monoclonal | |
1 mg/mL | |
ELISA 1:100 - 1:2000, Immunohistochemistry 10 - 20 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 10 - 20 ug/mL | |
P21359 | |
NF1 | |
A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] | |
0.2 mL | |
Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
4763 | |
Human, Mouse, Rat, Guinea Pig | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
McNFn27b | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human and rodent. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Neurofibromin 1 Antibody (McNFn27b) - BSA Free, Novus Biologicals™ > 0.2mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction