Learn More
Neurofilaments 70/200 kDa Monoclonal Antibody (2F11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515234
289.34 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
MA5-15234 targets Neurofilament (200kDa and 68kDa) in IF, IHC (P), IP, and WB applications and shows reactivity with Feline, Human, mouse, Rabbit, and Rat samples. The MA5-15234 immunogen is neurofilament purified from human brain.
Neurofilaments usually contain three intermediate filament proteins: NEFL, NEFM, and NEFH which are involved in the maintenance of neuronal caliber. NEFH has an important function in mature axons that is not subserved by the two smaller NF proteins. May additionally cooperate with the neuronal intermediate filament proteins PRPH and INA to form neuronal filamentous networks.
Specifications
Neurofilaments 70/200 kDa | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 15mM sodium azide | |
P07196, P08551, P12036, P16884, P19246, P19527 | |
NEFH, NEFL | |
Neurofilament purified from human brain | |
RUO | |
100353082, 100357177, 101081039, 101087272, 18039, 24587, 380684, 4744, 4747, 83613 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
2F11 | |
Unconjugated | |
NEFH | |
200 kDa neurofilament protein; 68 kDa neurofilament protein; AI847934; CMT1F; CMT2CC; CMT2E; KIAA0845; light molecular weight neurofilament protein; LOW QUALITY PROTEIN: neurofilament light polypeptide; mKIAA0845; NEFH; Nefl; neurofilament 200kDa; neurofilament heavy; neurofilament heavy polypeptide; neurofilament light; neurofilament light polypeptide; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet H protein; Neurofilament triplet L protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament-H; NF200; NF68; NFH; NF-H; NFL; NF-L; PPP1R110; protein phosphatase 1, regulatory subunit 110 | |
Mouse | |
500 μL | |
Primary | |
Feline, Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG1 κ |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
More Discovery, Less Cost
Now extended - offer ends 28 MarchPromo Code: 24111Expires: 2025-03-28
Days
0Hours
16Minutes
39Seconds
53New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.